2,3,5,6-Tetrafluorthiophenol

Lieferant: FLUOROCHEM

Synonyme: 2,3,5,6-Tetrafluorbenzolthiol, 2,3,5,6-Tetrafluorphenylmercaptan, 1-Mercapto-2,3,5,6-tetrafluorbenzol, 2,3,5,6-Tetrafluorbenzol-1-thiol

010547-25G 010547-5G
FLUO010547-25GEA 0 EUR
FLUO010547-25G FLUO010547-5G
2,3,5,6-Tetrafluorthiophenol
2,3,5,6-Tetrafluorthiophenol
Formel: C₆H₂F₄S
Molecular Weight: 182.14 g/mol
Siedepunkt: 152
MDL: MFCD00004829
CAS-Nummer: 769-40-4
EINECS: 212-210-1
UN: 1992
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR