2,3,4-Trifluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,3,4-Trifluorbenzol-1-sulfonylchlorid, 2,3,4-Trifluorbenzolsulfochlorid, 2,3,4-Trifluorbenzolsulfonsäurechlorid

CFF685.5 007483-5G 007483-25G
CHMPCFF685.5EA 532 EUR
CHMPCFF685.5 FLUO007483-5G FLUO007483-25G
2,3,4-Trifluorbenzolsulfonylchlorid
Trifluorobenzenesulphonyl chloride
Formel: C₆H₂ClF₃O₂S
Molecular Weight: 230,59 g/mol
Siedepunkt: 59…61 °C (0,1 mmHg)
Dichte: 1,64 g/cm³ (20 °C)
MDL: MFCD00102573
CAS-Nummer: 175278-08-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR