2,4,5-Trifluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

CFF1273. 005518-5G 005518-1G
CHMPCFF1273.EA 0 EUR
CHMPCFF1273. FLUO005518-5G FLUO005518-1G
2,4,5-Trifluorbenzolsulfonylchlorid
2,4,5-Trifluorbenzolsulfonylchlorid
Formel: C₆H₂ClF₃O₂S
MDL: MFCD00077600
CAS-Nummer: 220227-21-4
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR