2,4-Difluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,4-Difluorbenzolsulfonsäurechlorid, 2,4-Difluorbenzol-1-sulfonylchlorid, 2,4-Difluorbenzolsulfochlorid

CFF611.5 003579-5G 003579-10G 003579-25G
CHMPCFF611.5EA 508 EUR
CHMPCFF611.5 FLUO003579-5G FLUO003579-10G FLUO003579-25G
2,4-Difluorbenzolsulfonylchlorid
2,4-Difluorbenzolsulfonylchlorid
Formel: C₆H₃ClF₂O₂S
Molecular Weight: 212.6 g/mol
Siedepunkt: 84
Dichte: 1.581 g/cm³ (20 °C)
MDL: MFCD00042481
CAS-Nummer: 13918-92-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR