2,5-Difluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,5-Difluorbenzol-1-sulfonylchlorid, 2,5-Difluorbenzolsulfonsäurechlorid, 2,5-Difluorbenzolsulfochlorid

CFF1020.1 005515-1G 005515-5G 005515-25G
CHMPCFF1020.1EA 522 EUR
CHMPCFF1020.1 FLUO005515-1G FLUO005515-5G FLUO005515-25G
2,5-Difluorbenzolsulfonylchlorid
2,5-Difluorbenzolsulfonylchlorid
Formel: C₆H₃ClF₂O₂S
Molecular Weight: 212.6 g/mol
MDL: MFCD00060682
CAS-Nummer: 26120-86-5
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR