2,5-Difluorbenzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,5-Difluorbenzol-1-sulfonylchlorid, 2,5-Difluorbenzolsulfonsäurechlorid, 2,5-Difluorbenzolsulfochlorid
M-Clarity™ Qualitätsstufe = MQ200
555428-1G
555428-5G
SIAL555428-1GEA
88.3
EUR
SIAL555428-1G
SIAL555428-5G
2,5-Difluorbenzolsulfonylchlorid, Sigma-Aldrich®
2,5-Difluorbenzolsulfonylchlorid
Formel:
C₆H₃ClF₂O₂S Molecular Weight: 212,6 g/mol |
MDL:
MFCD00060682 CAS-Nummer: 26120-86-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...