2,6-Difluorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2,6-Difluorbenzolsulfonsäurechlorid, 2,6-Difluorbenzol-1-sulfonylchlorid, 2,6-Difluorbenzolsulfochlorid
CFF203.5
CFF203.10
004780-1G
004780-5G
004780-25G
CHMPCFF203.5EA
268
EUR
CHMPCFF203.5
CHMPCFF203.10
FLUO004780-1G
FLUO004780-5G
FLUO004780-25G
2,6-Difluorbenzolsulfonylchlorid
2,6-Difluorbenzolsulfonylchlorid
Formel:
C₆H₃ClF₂O₂S Molecular Weight: 212.6 g/mol Siedepunkt: 127.5…129.5 °C (1013 hPa) Dichte: 1.568 g/cm³ (20 °C) |
MDL:
MFCD00085004 CAS-Nummer: 60230-36-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...