3,4-Difluorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3,4-Difluorbenzolsulfochlorid, 3,4-Difluorbenzol-1-sulfonylchlorid, 3,4-Difluorbenzolsulfonsäurechlorid
CFF612.5
004781-5G
004781-25G
CHMPCFF612.5EA
508
EUR
CHMPCFF612.5
FLUO004781-5G
FLUO004781-25G
3,4-Difluorbenzolsulfonylchlorid
3,4-Difluorbenzolsulfonylchlorid
Formel:
C₆H₃ClF₂O₂S Siedepunkt: 105 °C (15 mmHg) Dichte: 1,586 g/cm³ (20 °C) |
MDL:
MFCD00153120 CAS-Nummer: 145758-05-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...