3,5-Difluorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3,5-Difluorbenzolsulfonsäurechlorid, 3,5-Difluorbenzol-1-sulfonylchlorid, 3,5-Difluorbenzolsulfochlorid
013068-5G
013068-25G
FLUO013068-5GEA
32
EUR
FLUO013068-5G
FLUO013068-25G
3,5-Difluorbenzolsulfonylchlorid
3,5-Difluorbenzolsulfonylchlorid
Formel:
C₆H₃ClF₂O₂S Molecular Weight: 212.6 g/mol |
MDL:
MFCD02091378 CAS-Nummer: 210532-25-5 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...