3,5-Difluorbenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Difluorbenzolsulfonsäurechlorid, 3,5-Difluorbenzol-1-sulfonylchlorid, 3,5-Difluorbenzolsulfochlorid

M-Clarity™ Qualitätsstufe = MQ100

555975-1G 555975-5G
SIAL555975-1GEA 26 EUR
SIAL555975-1G SIAL555975-5G
3,5-Difluorbenzolsulfonylchlorid, Sigma-Aldrich®
3,5-Difluorbenzolsulfonylchlorid
Formel: C₆H₃ClF₂O₂S
Molecular Weight: 212,6 g/mol
MDL: MFCD02091378
CAS-Nummer: 210532-25-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR