3-Fluorbenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 3-Fluorbenzolsulfonsäurechlorid, 3-Fluorbenzolsulfochlorid, 3-Fluorbenzol-1-sulfonylchlorid

TL00519DA TL00519EA TL00519.50 TL00519EB
MAYBTL00519DAEA 0 EUR
MAYBTL00519DA MAYBTL00519EA MAYBTL00519.50 MAYBTL00519EB
3-Fluorbenzolsulfonylchlorid
3-Fluorbenzolsulfonylchlorid
Formel: C₆H₄ClFO₂S
Molecular Weight: 194,61 g/mol
MDL: MFCD00042286
CAS-Nummer: 701-27-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR