3-Fluorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
CFF262.10
CFF262.50
003577-5G
003577-25G
CHMPCFF262.10EA
321
EUR
CHMPCFF262.10
CHMPCFF262.50
FLUO003577-5G
FLUO003577-25G
3-Fluorbenzolsulfonylchlorid
3-Fluorbenzolsulfonylchlorid
Formel:
C₆H₄ClFO₂S |
MDL:
MFCD00042286 CAS-Nummer: 701-27-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...