3-Fluorbenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

532738-1G 532738-5G
SIAL532738-1GEA 18.3 EUR
SIAL532738-1G SIAL532738-5G
3-Fluorbenzolsulfonylchlorid, Sigma-Aldrich®
3-Fluorbenzolsulfonylchlorid
Formel: C₆H₄ClFO₂S
MDL: MFCD00042286
CAS-Nummer: 701-27-9
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR