Methyl-2-iodbenzoat

Lieferant: Thermo Scientific

Synonyme: 2-Iodbenzoesäuremethylester

CD04352.1 CD04352.10 CD04352.50
MAYBCD04352.1EA 0 EUR
MAYBCD04352.1 MAYBCD04352.10 MAYBCD04352.50
Methyl-2-iodbenzoat
Methyl-2-iodbenzoat
Formel: C₈H₇IO₂
Molecular Weight: 262,05 g/mol
Siedepunkt: 272 °C (1013 hPa)
Dichte: 1,784 g/cm³ (20 °C)
MDL: MFCD00016351
CAS-Nummer: 610-97-9
EINECS: 210-243-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR