Ethyl-4-iodbenzoat

Lieferant: FLUOROCHEM

Synonyme: 4-Iodbenzoesäureethylester

005465-25G 005465-100G
FLUO005465-25GEA 36.9 EUR
FLUO005465-25G FLUO005465-100G
Ethyl-4-iodbenzoat
Ethyl-4-iodbenzoat
Formel: C₉H₉IO₂
Molecular Weight: 276,07 g/mol
Siedepunkt: 281 °C (1013 hPa)
Dichte: 1,655 g/cm³ (20 °C)
MDL: MFCD00017344
CAS-Nummer: 51934-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR