2,5-Diiodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

107905-1G 107905-5G
SIAL107905-1GEA 72.3 EUR
SIAL107905-1G SIAL107905-5G
2,5-Diiodbenzoesäure, Sigma-Aldrich®
2,5-Diiodbenzoesäure
Formel: C₇H₄I₂O₂
Molecular Weight: 373,92 g/mol
MDL: MFCD00045801
CAS-Nummer: 14192-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR