3-Iodphenol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
I10007-25G
I10007-5G
SIALI10007-25GEA
79.7
EUR
SIALI10007-25G
SIALI10007-5G
3-Iodphenol, Sigma-Aldrich®
3-Iodphenol
Formel:
C₆H₅IO Molecular Weight: 220.01 g/mol |
MDL:
MFCD00002261 CAS-Nummer: 626-02-8 EINECS: 210-923-2 UN: 3335 ADR: 9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...