N,N,N',N'-Tetramethyl-p-phenylendiamin ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: TMPD, 1,4-Bis(dimethylamino)benzol, N,N,N',N'-Tetramethyl-1,4-phenylendiamin
L00559.09 L00559.18
L00559.09EA 48.6 EUR
L00559.09 L00559.18
N,N,N',N'-Tetramethyl-p-phenylendiamin ≥98%
N,N,N',N'-Tetramethyl-p-phenylendiamin
Purity: 98+% CAS No.: 100-22-1 Molecular Formula: C10H16N2 Formula Weight: 164.25 Storage Temperature: 2°C to 8°C Melting Point: 48-52° Boiling Point: 260° Beilstein Reference: 1564025 Merck Reference: 14,9228 UN No.: 1673 MDL No.: MFCD00008309
Formel: C₁₀H₁₆N₂
Molecular Weight: 164.25 g/mol
Siedepunkt: 260 °C
Schmelzpunkt: 48…52 °C
Dichte: 1.08 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00008309
CAS-Nummer: 100-22-1
EINECS: 202-831-6
UN: 1673
ADR: 6.1 III
Merck Index: 12,09367
Bestellen
TECHNISCHE DATEN
Environmentally Preferable