3,5-Dimethoxyphenylisocyanat

Lieferant: CHEMPUR

SB01905.250 SB01905.1
CHMPSB01905.250EA 0 EUR
CHMPSB01905.250 MAYBSB01905.1
3,5-Dimethoxyphenylisocyanat
3,5-Dimethoxyphenylisocyanat
Formel: C₉H₉NO₃
Molecular Weight: 179,18 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 54132-76-2
UN: 3335
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR