2-Nitrophenylisothiocyanat

Lieferant: FLUOROCHEM

009549-1G 009549-5G
FLUO009549-1GEA 34.1 EUR
FLUO009549-1G FLUO009549-5G
2-Nitrophenylisothiocyanat
2-Nitrophenylisothiocyanat
Formel: C₇H₄N₂O₂S
Molecular Weight: 180.19 g/mol
Siedepunkt: 135…137 °C (17 mmHg)
Schmelzpunkt: 130…131 °C
MDL: MFCD00041238
CAS-Nummer: 2719-30-4
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR