3-Nitrophenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-Isothiocyanato-3-nitrobenzene, m-Nitrophenylisothiocyanat
009550-1G
009550-5G
FLUO009550-1GEA
0
EUR
FLUO009550-1G
FLUO009550-5G
3-Nitrophenylisothiocyanat
3-Nitrophenylisothiocyanat
Formel:
C₇H₄N₂O₂S Molecular Weight: 180,19 g/mol Siedepunkt: 275…280 °C (1013 hPa) Schmelzpunkt: 60,5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00041245 CAS-Nummer: 3529-82-6 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...