3,4-Dimethoxyphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1,2-Dimethoxy-4-isothiocyanatobenzol

008867-5G 008867-25G
FLUO008867-5GEA 0 EUR
FLUO008867-5G FLUO008867-25G
3,4-Dimethoxyphenylisothiocyanat
3,4-Dimethoxyphenylisothiocyanat
Formel: C₉H₉NO₂S
Molecular Weight: 195,24 g/mol
Siedepunkt: 171…172 °C (7 mmHg)
Schmelzpunkt: 48…48,5 °C
MDL: MFCD00041075
CAS-Nummer: 33904-04-0
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR