2,4-Dimethoxyphenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 1,3-Dimethoxy-4-isothiocyanatobenzol

SB01848.1 SB01848.10 SB01848.50
MAYBSB01848.1EA 0 EUR
MAYBSB01848.1 MAYBSB01848.10 MAYBSB01848.50
2,4-Dimethoxyphenylisothiocyanat
2,4-Dimethoxyphenylisothiocyanat
Formel: C₉H₉NO₂S
Molecular Weight: 195,24 g/mol
Siedepunkt: 166…167 °C (6 mmHg)
Schmelzpunkt: 57 °C
MDL: MFCD00041058
CAS-Nummer: 33904-03-9
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR