2,4-Dimethoxyphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1,3-Dimethoxy-4-isothiocyanatobenzol
008858-5G
008858-25G
FLUO008858-5GEA
107
EUR
FLUO008858-5G
FLUO008858-25G
2,4-Dimethoxyphenylisothiocyanat
2,4-Dimethoxyphenylisothiocyanat
Formel:
C₉H₉NO₂S Molecular Weight: 195.24 g/mol Siedepunkt: 166…167 °C (6 mmHg) Schmelzpunkt: 57 |
MDL:
MFCD00041058 CAS-Nummer: 33904-03-9 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...