3,5-Dimethoxyphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1,3-Dimethoxy-5-isothiocyanatobenzene
008921-1G
008921-5G
008921-25G
FLUO008921-1GEA
72.9
EUR
FLUO008921-1G
FLUO008921-5G
FLUO008921-25G
3,5-Dimethoxyphenylisothiocyanat
3,5-Dimethoxyphenylisothiocyanat
Formel:
C₉H₉NO₂S Siedepunkt: 130…132 °C (3 mmHg) Schmelzpunkt: 51 °C |
MDL:
MFCD00041077 CAS-Nummer: 104968-58-3 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...