2,5-Dimethoxyphenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 1,4-Dimethoxy-2-isothiocyanatobenzene

SB01666.1 SB01666EA SB01666.50
MAYBSB01666.1EA 0 EUR
MAYBSB01666.1 MAYBSB01666EA MAYBSB01666.50
2,5-Dimethoxyphenylisothiocyanat
2,5-Dimethoxyphenylisothiocyanat
Formel: C₉H₉NO₂S
Molecular Weight: 195,24 g/mol
Siedepunkt: 178…180 °C (16 mmHg)
Schmelzpunkt: 33 °C
MDL: MFCD00041059
CAS-Nummer: 40532-06-7
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR