2,5-Dimethoxyphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1,4-Dimethoxy-2-isothiocyanatobenzene
008866-5G
008866-25G
FLUO008866-5GEA
116
EUR
FLUO008866-5G
FLUO008866-25G
2,5-Dimethoxyphenylisothiocyanat
2,5-Dimethoxyphenylisothiocyanat
Formel:
C₉H₉NO₂S Molecular Weight: 195,24 g/mol Siedepunkt: 178…180 °C (16 mmHg) Schmelzpunkt: 33 °C |
MDL:
MFCD00041059 CAS-Nummer: 40532-06-7 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...