2-(Methylthio)phenylisothiocyanat

Lieferant: FLUOROCHEM

009542-1G 009542-5G 009542-25G
FLUO009542-1GEA 51.8 EUR
FLUO009542-1G FLUO009542-5G FLUO009542-25G
2-(Methylthio)phenylisothiocyanat
2-(Methylthio)phenylisothiocyanat
Formel: C₈H₇NS₂
Molecular Weight: 181.28 g/mol
Siedepunkt: 270
Dichte: 1.227 g/cm³ (20 °C)
MDL: MFCD00041061
CAS-Nummer: 51333-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR