1,2-Phenylendiisothiocyanat

Lieferant: FLUOROCHEM

009564-1G 009564-5G
FLUO009564-1GEA 76.2 EUR
FLUO009564-1G FLUO009564-5G
1,2-Phenylendiisothiocyanat
1,2-Phenylendiisothiocyanat
Formel: C₈H₄N₂S₂
MDL: MFCD00060366
CAS-Nummer: 71105-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR