2-Amino-3-nitrophenol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-6-nitroanilin
382040050
382040250
ACRO382040050EA
39.2
EUR
ACRO382040050
ACRO382040250
2-Amino-3-nitrophenol 98%
2-Amino-3-nitrophenol
Formel:
C₆H₆N₂O₃ Molecular Weight: 154.13 g/mol Schmelzpunkt: 215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010875 CAS-Nummer: 603-85-0 EINECS: 210-060-1 |
Spezifikation Testergebnisse
Appearance | Red-brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...