2-Amino-3-nitrophenol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxy-6-nitroanilin

H27472.14 H27472.06
H27472.14EA 234 EUR
H27472.14 H27472.06
2-Amino-3-nitrophenol 98%
2-Amino-3-nitrophenol

Purity: 98% CAS No.: 603-85-0 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: ca 212° Beilstein Reference: 2804307

Formel: C₆H₆N₂O₃
Molecular Weight: 154.13 g/mol
Schmelzpunkt: ∼ 212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010875
CAS-Nummer: 603-85-0
EINECS: 210-060-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR