(2-Aminoethoxy)benzol 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: β-Aminophenetol, (2-Aminoethyl)phenylether, 2-Phenoxyethylamin, ω-Aminophenetol

A12225.14 A12225.03 A12225.06
A12225.14EA 461 EUR
A12225.14 A12225.03 A12225.06
(2-Aminoethoxy)benzol 98%
(2-Aminoethoxy)benzol

Purity: 98% CAS No.: 1758-46-9 Molecular Formula: C8H11NO Formula Weight: 137.18 Flash Point: 107°(224°F) Refractive Index: 1.5370 Odour: Amine-like Boiling Point: 88-90°/1mm Beilstein Reference: 774671 UN No.: 2735 MDL No.: MFCD00052975

Formel: C₆H₅OCH₂CH₂NH₂
Molecular Weight: 137,18 g/mol
Siedepunkt: 88…90 °C (1 mmHg)
Dichte: 1,048
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052975
CAS-Nummer: 1758-46-9
EINECS: 217-153-6
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR