2-Aminophenol 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxyanilin

A13735.22 A13735.36
A13735.22EA 30.5 EUR
A13735.22 A13735.36
2-Aminophenol 99%
2-Aminophenol

Purity: 99% CAS No.: 95-55-6 Molecular Formula: C6H7NO Formula Weight: 109.13 Flash Point: 175°(347°F) Odour: Characteristic Boiling Point: 150°/11mm Beilstein Reference: 606075 Merck Reference: 14,461 UN No.: 2512 MDL No.: MFCD00007690

Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
Siedepunkt: 150 °C (11 mmHg)
Schmelzpunkt: 173…177 °C
Dichte: 1,328
Flammpunkt: >175 °C (347 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007690
CAS-Nummer: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 14,461

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR