3-Aminophenol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Hydroxyanilin
A11943.0B
A11943.0I
A11943.30
A11943.0BEA
227
EUR
A11943.0B
A11943.0I
A11943.30
3-Aminophenol ≥98%
3-Aminophenol
Purity: 98+%
CAS No.: 591-27-5
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 178°(352°F)
Odour: Characteristic
pH: 6.8
Melting Point: 122-126°
Boiling Point: 164°/11mm
Beilstein Reference: 636059
Merck Reference: 14,460
UN No.: 2512
MDL No.: MFCD00007786
Formel:
C₆H₇NO Molecular Weight: 109,13 g/mol Siedepunkt: 164 °C (11 mmHg) Schmelzpunkt: 122…126 °C Dichte: 0,99 Flammpunkt: 178 °C (352 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007786 CAS-Nummer: 591-27-5 EINECS: 209-711-2 UN: 2512 ADR: 6.1,III Merck Index: 14,460 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...