4-Aminophenol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Hydroxyanilin

A13581.30 A13581.0B A13581.0I A13581.18
A13581.30EA 37.3 EUR
A13581.30 A13581.0B A13581.0I A13581.18
4-Aminophenol 98%
4-Aminophenol

Purity: 98% CAS No.: 123-30-8 Molecular Formula: C6H7NO Formula Weight: 109.13 Flash Point: 195°(383°F) Physical Form: Powder Density: 1.29 Sensitivity: Air and Light Sensitive Appearance: White, pale brown/purple Odour: Phenol-like Melting Point: 186-190° Boiling Point: 284° dec. Beilstein Reference: 385836 Merck Reference: 14,462 UN No.: 2512 MDL No.: MFCD00007869

Formel: H₂NC₆H₄OH
Molecular Weight: 109,13 g/mol
Siedepunkt: 284 °C (Zersetzung)
Schmelzpunkt: 186…190 °C
Dichte: 1,29
Flammpunkt: 195 °C (383 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007869
CAS-Nummer: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III
Merck Index: 14,462
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7af17b754d4ebaa0993b4544d4e6ab6c116724c&vid=6275667c2624ee686f61715acd420dc596e076fa&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR