Sodium sulphanilate dihydrate sodium salt, Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
4-Aminobenzenesulphonic acid, sodium salt dihydrate
0215671683
0215671650
ICNA0215671683EA
90.5
EUR
ICNA0215671683
ICNA0215671650
Sodium sulphanilate dihydrate sodium salt, Pulver
Sodium sulfanilate dihydrate
Sulphanilic acid is a reagent for chromatographic detection of histidine, and intermediate for synthesis of dyes and pharmaceuticals.
Formel:
C₆H₁₀NNaO₅S Molecular Weight: 231,198 g/mol Siedepunkt: 363 °C (1013 hPa) Schmelzpunkt: 279 °C Dichte: 1,485 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064394 CAS-Nummer: 6106-22-5 EINECS: 204-482-5 Merck Index: 13,09012 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...