p-Phenylendiamin 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: PPD, 1,4-Diaminobenzol, 1,4-Phenylendiamin

130575000 130570050 130570025
ACRO130575000EA 146 EUR
ACRO130575000 ACRO130570050 ACRO130570025
p-Phenylendiamin 99+%
p-Phenylendiamin
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
Siedepunkt: 267
Schmelzpunkt: 139…141 °C
Flammpunkt: 156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007901
CAS-Nummer: 106-50-3
EINECS: 203-404-7
UN: 1673
ADR: 6.1 III
Merck Index: 12,07439

Bestellen

Spezifikation Testergebnisse

Appearance White to grey to brown to purple Powder or flakes (May darken on storage)
Infrared spectrum Conforms
GC ≥99.0 %
HPLC ≥96.0 %
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR