p-Phenylendiamin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: PPD, 1,4-Diaminobenzol, 1,4-Phenylendiamin

A15680.0B A15680.0I A15680.30
A15680.0BEA 103 EUR
A15680.0B A15680.0I A15680.30
p-Phenylendiamin 97%
p-Phenylendiamin

Purity: 97% CAS No.: 106-50-3 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 156°(312°F) Physical Form: Cryst solid Odour: Weak Melting Point: 138-142° Boiling Point: 267-271° Solubility: Soluble in alcohol, chloroform, ether Beilstein Reference: 742029 Merck Reference: 14,7285 UN No.: 1673 MDL No.: MFCD00007901

Formel: C₆H₈N₂
Molecular Weight: 108,14 g/mol
Siedepunkt: 267…271 °C
Schmelzpunkt: 138…142 °C
Dichte: 1,251 g/cm³ (20 °C)
Flammpunkt: 156 °C (312 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007901
CAS-Nummer: 106-50-3
EINECS: 203-404-7
UN: 1673
ADR: 6.1,III
Merck Index: 14,7285

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR