3-Nitro-o-phenylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine
M-Clarity™ Qualitätsstufe = MQ100
N21308-1G
N21308-5G
SIALN21308-1GEA
58.4
EUR
SIALN21308-1G
SIALN21308-5G
3-Nitro-o-phenylendiamin, Sigma-Aldrich®
3-Nitro-o-phenylendiamin
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,14 g/mol Schmelzpunkt: 158…159 °C |
MDL:
MFCD00007722 CAS-Nummer: 3694-52-8 EINECS: 223-013-5 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...