4-Nitro-o-phenylendiamin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine
148841000
148845000
ACRO148841000EA
79.8
EUR
ACRO148841000
ACRO148845000
4-Nitro-o-phenylendiamin 98%
4-Nitro-o-phenylendiamin
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,14 g/mol Schmelzpunkt: 196…201 °C |
MDL:
MFCD00007724 CAS-Nummer: 99-56-9 EINECS: 202-766-3 Merck Index: 12,06720 |
Spezifikation Testergebnisse
Appearance | Red-brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...