2-Nitro-p-phenylendiamin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Nitro-1,4-phenylenediamine, 2-Nitrobenzene-1,4-diamine

B23477.22 B23477.14 B23477.36
B23477.22EA 67 EUR
B23477.22 B23477.14 B23477.36
2-Nitro-p-phenylendiamin 95%
2-Nitro-p-phenylendiamin

Purity: 95% CAS No.: 5307-14-2 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 134-137° MDL No.: MFCD00007903

Formel: C₆H₇N₃O₂
Molecular Weight: 153.14 g/mol
Schmelzpunkt: 134…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007903
CAS-Nummer: 5307-14-2
EINECS: 226-164-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR