Bariumdiphenylamin-4-sulfonat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bariumbis(4-anilinobenzolsulfonat), 4-(Phenylamino)benzolsulfonsäure Bariumsalz, Barium-N-phenylsulfanilat, Diphenylamin-4-sulfonsäure Bariumsalz
A16197.06
A16197.14
A16197.06EA
47.4
EUR
A16197.06
A16197.14
Bariumdiphenylamin-4-sulfonat
Bariumdiphenylamin-4-sulfonat
CAS No.: 6211-24-1
Molecular Formula: C24H20BaN2O6S2
Formula Weight: 633.90
Melting Point:
Formel:
C₂₄H₂₀BaN₂O₆S₂ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007497 CAS-Nummer: 6211-24-1 EINECS: 228-278-0 UN: 1564 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...