3-Nitrobenzolsulfonylchlorid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Nitrobenzolsulfochlorid, 3-Nitrobenzolsulfonsäurechlorid, 3-Nitrobenzol-1-sulfonylchlorid

415840250 415841000
ACRO415840250EA 72.8 EUR
ACRO415840250 ACRO415841000
3-Nitrobenzolsulfonylchlorid 97%
3-Nitrobenzolsulfonylchlorid
Formel: C₆H₄ClNO₄S
Molecular Weight: 221.62 g/mol
Schmelzpunkt: 60…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007435
CAS-Nummer: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange light beige Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 64 °C
HPLC ≥96.0 %
Specific optical rotation -105° to -109° (20 °C, 589 nm) (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR