Benzoesäureanhydrid ≥98%

Lieferant: Thermo Scientific
Achtung

A14269.0B A14269.18 A14269.30
A14269.0BEA 228 EUR
A14269.0B A14269.18 A14269.30
Benzoesäureanhydrid ≥98%
Benzoic anhydride

Purity: 98% CAS No.: 93-97-0 Molecular Formula: C14H10O3 Formula Weight: 226.23 Flash Point: 110°(230°F) Physical Form: Powder Density: 1.199 Sensitivity: Moisture Sensitive Odour: Odourless Melting Point: 40-44° Boiling Point: 360° Beilstein Reference: 516726 Merck Reference: 14,1092 MDL No.: MFCD00003073

Formel: C₁₄H₁₀O₃
Molecular Weight: 226,23 g/mol
Siedepunkt: 360 °C
Schmelzpunkt: 40…44 °C
Dichte: 1,199
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003073
CAS-Nummer: 93-97-0
EINECS: 202-291-1
Merck Index: 14,1092

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR