2-(Methylthio)phenol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-Hydroxythioanisole
001770-5G
001770-25G
FLUO001770-5GEA
60.9
EUR
FLUO001770-5G
FLUO001770-25G
2-(Methylthio)phenol
2-(Methylthio)phenol
Formel:
C₇H₈OS Molecular Weight: 140.21 g/mol Siedepunkt: 81…82 °C (0.5 mmHg) Dichte: 1.168 g/cm³ (25 °C) |
MDL:
MFCD00002211 CAS-Nummer: 1073-29-6 EINECS: 214-027-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...