1,4-Diphenoxybenzol ≥98%
Lieferant: Thermo Fisher Scientific
A14864.06
A14864.14
A14864.22
A14864.06EA
71.8
EUR
A14864.06
A14864.14
A14864.22
1,4-Diphenoxybenzol ≥98%
1,4-Diphenoxybenzene
Purity: 98%
CAS No.: 3061-36-7
Molecular Formula: C18H14O2
Formula Weight: 262.31
Flash Point: 202°(395°F)
Melting Point: 72-73°
Boiling Point: 172°/0.01mm
Beilstein Reference: 2215945
MDL No.: MFCD00038368
Formel:
C₁₈H₁₄O₂ Siedepunkt: 172 °C (0,01 mmHg) Schmelzpunkt: 72…73 °C Dichte: 1,083 Flammpunkt: 202 °C (395 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038368 CAS-Nummer: 3061-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...