1,4-Benzoldithiol ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Benzol-1,4-dithiol

L19440.03 L19440.06 L19440.MD
L19440.03EA 147 EUR
L19440.03 L19440.06 L19440.MD
1,4-Benzoldithiol ≥97%
1,4-Benzoldithiol

Purity: 97% CAS No.: 624-39-5 Molecular Formula: C6H6S2 Formula Weight: 142.24 Storage Temperature: 2°C to 8°C Odour: Stench Beilstein Reference: 1680022 UN No.: 3335 MDL No.: MFCD00142541

Formel: C₆H₆S₂
Siedepunkt: 240 °C (1013 hPa)
Schmelzpunkt: 95…99 °C
MDL: MFCD00142541
CAS-Nummer: 624-39-5
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR