trans-Chalkon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: trans-Benzylidenacetophenon, trans-1,3-Diphenyl-3-propen-1-on, trans-Benzalacetophenon

A14734.22 A14734.36
A14734.22EA 43.2 EUR
A14734.22 A14734.36
trans-Chalkon ≥97%
trans-Chalkon

Purity: 97% CAS No.: 614-47-1 Molecular Formula: C15H12O Formula Weight: 208.26 Melting Point: 55-57° Boiling Point: 208°/25mm Merck Reference: 14,2037 MDL No.: MFCD00003082

Formel: C₁₅H₁₂O
Siedepunkt: 208 °C (25 mmHg)
Schmelzpunkt: 55…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003082
CAS-Nummer: 614-47-1
EINECS: 210-383-8
Merck Index: 14,2037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f708bafe7d3c2e194d3e9bf5f410a2c30afb756&vid=aa90a6b415f359c4551772765e81cbdff2cce599&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR