trans-Stilben ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: trans-1,2-Diphenylethen, trans-1,2-Diphenylethylen

A10232.0E A10232.22 A10232.36
A10232.0EEA 1417 EUR
A10232.0E A10232.22 A10232.36
trans-Stilben ≥98%
trans-Stilben

Purity: 98% CAS No.: 103-30-0 Molecular Formula: C14H12 Formula Weight: 180.25 Odour: Odourless Melting Point: 122-125° Boiling Point: 305-307° Beilstein Reference: 1616740 Merck Reference: 14,8817 UN No.: 3077 MDL No.: MFCD00064300

Formel: C₁₄H₁₂
Molecular Weight: 180.25 g/mol
Siedepunkt: 305…307 °C
Schmelzpunkt: 122…125 °C
Dichte: 1.14
Lagertemperatur: Raumtemperatur
MDL: MFCD00064300
CAS-Nummer: 103-30-0
EINECS: 203-098-5
ADR: 9 III
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR