1-Brom-4-ethinylbenzol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Ethinyl-4-brombenzol, (4-Bromphenyl)acetylen, (4-Bromphenyl)ethin
L19082.03
L19082.MD
L19082.03EA
74.1
EUR
L19082.03
L19082.MD
1-Brom-4-ethinylbenzol ≥97%
1-Brom-4-ethinylbenzol
Purity: 97%
CAS No.: 766-96-1
Molecular Formula: C8H5Br
Formula Weight: 181.04
Odour: Odourless
Melting Point: 64-67°
Boiling Point: 88-90°/16mm
Beilstein Reference: 1209726
MDL No.: MFCD00168822
Formel:
C₈H₅Br Molecular Weight: 181,03 g/mol Siedepunkt: 88…90 °C (16 mmHg) Schmelzpunkt: 64…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168822 CAS-Nummer: 766-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...